COMH42122385 Combi-Blocks, Inc.

2-Azabicyclo[2.1.1]hexane-1-carboxylic acid hydrochloride

  • MDL number MFCD08460278
  • Cas Number : 116129-07-8
  • Molecular Weight : 163.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6649

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6649
Availability details for
Quantity
  • Availability
  • Add to Cart