COMH42122398 Combi-Blocks, Inc.

Methyl 2-(2-(bromomethyl)phenyl)-2-(methoxyimino)acetate

  • MDL number MFCD16038451
  • Cas Number : 115199-26-3
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9904

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9904
Availability details for
Quantity
  • Availability
  • Add to Cart