COMH42122399 Combi-Blocks, Inc.

Cbz-nh-peg(20)-cooh

  • MDL number MFCD21363333
  • Cas Number : 1334177-88-6
  • Molecular Weight : 1104.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9966
Availability details for
Quantity
  • Availability
  • Add to Cart