COMH421223A6 Combi-Blocks, Inc.

1-tert-Butyl 2-methyl 3-oxopiperidine-1,2-dicarboxylate

  • MDL number MFCD15071869
  • Cas Number : 122019-53-8
  • Molecular Weight : 257.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-0141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-0141
Availability details for
Quantity
  • Availability
  • Add to Cart