COMH421223C8 Combi-Blocks, Inc.

4-(4-Bromo-1h-pyrazol-1-yl)-1-ethylpiperidine

  • MDL number MFCD25957748
  • Cas Number : 1309961-03-2
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-0754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-0754
Availability details for
Quantity
  • Availability
  • Add to Cart