COMH421223D6 Combi-Blocks, Inc.

5,6-Dihydrobenzofuran-7(4h)-one

  • MDL number MFCD13184225
  • Cas Number : 108153-93-1
  • Molecular Weight : 136.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-0982

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-0982
Availability details for
Quantity
  • Availability
  • Add to Cart