COMH421223E4 Combi-Blocks, Inc.

4-Bromo-1h-indene

  • MDL number MFCD28158321
  • Molecular Weight : 195.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1108
Availability details for
Quantity
  • Availability
  • Add to Cart