COMH421223F8 Combi-Blocks, Inc.

4-(4-Formyl-phenoxy)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD09909952
  • Cas Number : 207798-38-7
  • Molecular Weight : 305.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1557
Availability details for
Quantity
  • Availability
  • Add to Cart