COMH4212241C Combi-Blocks, Inc.

N-Boc-peg24-alcohol

  • MDL number MFCD29052164
  • Molecular Weight : 1174.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-2211

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-2211
Availability details for
Quantity
  • Availability
  • Add to Cart