COMH42122444 Combi-Blocks, Inc.

4-(1H-1,2,3-Triazol-1yl)-benzonitrile

  • MDL number MFCD11100734
  • Cas Number : 85862-71-1
  • Molecular Weight : 170.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-2929

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-2929
Availability details for
Quantity
  • Availability
  • Add to Cart