COMH42122449 Combi-Blocks, Inc.

N-Boc-peg16-alcohol

  • MDL number MFCD29052165
  • Molecular Weight : 822.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3031
Availability details for
Quantity
  • Availability
  • Add to Cart