COMH42122476 Combi-Blocks, Inc.

1-Benzyl-3-methylpyrrolidin-3-amine

  • MDL number MFCD08273914
  • Cas Number : 181114-76-1
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-4170

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-4170
Availability details for
Quantity
  • Availability
  • Add to Cart