COMH42122489 Combi-Blocks, Inc.

Peg15

  • MDL number MFCD29079393
  • Cas Number : 28821-35-4
  • Molecular Weight : 678.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-4492

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-4492
Availability details for
Quantity
  • Availability
  • Add to Cart