COMH421224A7 Combi-Blocks, Inc.

Peg4-bis-nitrophenyl carbonate

  • MDL number MFCD29079397
  • Molecular Weight : 524.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5211

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5211
Availability details for
Quantity
  • Availability
  • Add to Cart