COMH421224B4 Combi-Blocks, Inc.

1-Isothiocyanato-peg3-t-butyl ester

  • MDL number MFCD29052177
  • Cas Number : 1835759-83-5
  • Molecular Weight : 319.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5447
Availability details for
Quantity
  • Availability
  • Add to Cart