COMH421224F5 Combi-Blocks, Inc.

5-Boc-4,5,6,7-tetrahydrofuro[3,2-c]pyridine

  • MDL number MFCD22574995
  • Cas Number : 179060-34-5
  • Molecular Weight : 223.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6801
Availability details for
Quantity
  • Availability
  • Add to Cart