COMH42122503 Combi-Blocks, Inc.

Iodocholine iodide

  • MDL number MFCD20489393
  • Cas Number : 28508-22-7
  • Molecular Weight : 356.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7033
Availability details for
Quantity
  • Availability
  • Add to Cart