COMH4212251C Combi-Blocks, Inc.

Propargyl-peg4-thiol

  • MDL number MFCD29764343
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7618
Availability details for
Quantity
  • Availability
  • Add to Cart