COMH4212251D Combi-Blocks, Inc.

Peg2-bis-isothiocyanato

  • MDL number MFCD27965508
  • Cas Number : 103144-37-2
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7647
Availability details for
Quantity
  • Availability
  • Add to Cart