COMH42122529 Combi-Blocks, Inc.

1-(2-Bromophenyl)-4-methylpiperazine

  • MDL number MFCD20483678
  • Cas Number : 250383-81-4
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7767
Availability details for
Quantity
  • Availability
  • Add to Cart