COMH42122544 Combi-Blocks, Inc.

6-Chloro-1h-benzo[d]imidazol-2-amine

  • MDL number MFCD08234599
  • Cas Number : 5418-93-9
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8296

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8296
Availability details for
Quantity
  • Availability
  • Add to Cart