COMH42122554 Combi-Blocks, Inc.

2-Hydroxy-2-(pyridin-4-yl)propanenitrile

  • MDL number MFCD00972010
  • Cas Number : 89976-64-7
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8652

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8652
Availability details for
Quantity
  • Availability
  • Add to Cart