COMH42122568 Combi-Blocks, Inc.

Tetra(t-butoxycarbonylethoxymethyl)methane

  • MDL number MFCD29042363
  • Cas Number : 175724-29-5
  • Molecular Weight : 648.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8984

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8984
Availability details for
Quantity
  • Availability
  • Add to Cart