COMH4212256C Combi-Blocks, Inc.

2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine

  • MDL number MFCD29089341
  • Cas Number : 851030-18-7
  • Molecular Weight : 327.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9030
Availability details for
Quantity
  • Availability
  • Add to Cart