COMH42122579 Combi-Blocks, Inc.

7-Aminomethyl-6-oxa-2-aza-spiro[3.4]octane-2-carboxylic acid tert-butyl ester

  • MDL number MFCD14581238
  • Cas Number : 1251002-26-2
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9248
Availability details for
Quantity
  • Availability
  • Add to Cart