COMH42122584 Combi-Blocks, Inc.

4-Nitrophenyl 3,6,9,12-tetraoxapentadec-14-ynyl carbonate

  • MDL number MFCD29041851
  • Cas Number : 1422540-81-5
  • Molecular Weight : 397.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9447
Availability details for
Quantity
  • Availability
  • Add to Cart