COMH42122587 Combi-Blocks, Inc.

Mpeg3-s-peg1t-butyl ester

  • MDL number MFCD29764331
  • Cas Number : 2365419-97-0
  • Molecular Weight : 352.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9487
Availability details for
Quantity
  • Availability
  • Add to Cart