COMH421225A8 Combi-Blocks, Inc.

Aminoxy-peg3-methane

  • MDL number MFCD26910997
  • Cas Number : 248275-10-7
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9962

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9962
Availability details for
Quantity
  • Availability
  • Add to Cart