COMH421225BD Combi-Blocks, Inc.

Chloromethyl dodecanoate

  • MDL number MFCD29924218
  • Cas Number : 61413-67-0
  • Molecular Weight : 248.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0667

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0667
Availability details for
Quantity
  • Availability
  • Add to Cart