COMH421225C4 Combi-Blocks, Inc.

sodium (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(isopropylsulfanyl)oxane-2-carboxylate

  • MDL number MFCD00798401
  • Cas Number : 208589-93-9
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-0843
Availability details for
Quantity
  • Availability
  • Add to Cart