COMH421225D2 Combi-Blocks, Inc.

Ethyl 3-(4-ethoxyphenyl)-3-oxopropanoate

  • MDL number MFCD11193513
  • Cas Number : 51725-83-8
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1555

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1555
Availability details for
Quantity
  • Availability
  • Add to Cart