COMH421225D7 Combi-Blocks, Inc.

Fmoc-5,6-dehydrohomoleucine

  • MDL number MFCD30475879
  • Cas Number : 1369532-04-6
  • Molecular Weight : 365.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1683

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1683
Availability details for
Quantity
  • Availability
  • Add to Cart