COMH421225D8 Combi-Blocks, Inc.

nickel acetate

  • MDL number MFCD00012451
  • Cas Number : 373-02-4
  • Molecular Weight : 176.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1706

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1706
Availability details for
Quantity
  • Availability
  • Add to Cart