COMH421225DA Combi-Blocks, Inc.

Methyl 4-(cyclopropylmethoxy)-2-chlorobenzoate

  • MDL number MFCD24368740
  • Cas Number : 154606-03-8
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1777
Availability details for
Quantity
  • Availability
  • Add to Cart