COMH421225EB Combi-Blocks, Inc.

Ethyl 2-amino-1h-benzo[d]imidazole-5-carboxylate

  • MDL number MFCD08282516
  • Cas Number : 24370-20-5
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2194

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2194
Availability details for
Quantity
  • Availability
  • Add to Cart