COMH421225F6 Combi-Blocks, Inc.

2,4-dihydroxy-5-iodobenzoic acid

  • MDL number MFCD20257782
  • Cas Number : 856106-63-3
  • Molecular Weight : 280.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2603
Availability details for
Quantity
  • Availability
  • Add to Cart