COMH42122611 Combi-Blocks, Inc.

4-Iodo-2-isopropyl-1-methyl-1h-imidazole

  • MDL number MFCD29924215
  • Cas Number : 851870-24-1
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3636

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3636
Availability details for
Quantity
  • Availability
  • Add to Cart