COMH4212262B Combi-Blocks, Inc.

sulfuric acid methyl (1R,15R,18R)-17-ethyl-3,13-diazapentacyclo[13.3.1.0^{2,10}.0^{4,9}.0^{13,18}]nonadeca-2(10),4(9),5,7,16-pentaene-1-carboxylate

  • MDL number MFCD06410943
  • Cas Number : 70674-90-7
  • Molecular Weight : 434.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4138

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4138
Availability details for
Quantity
  • Availability
  • Add to Cart