COMH42122631 Combi-Blocks, Inc.

3-Hydroxy-2-trifluoromethylpropionic acid

  • MDL number MFCD00042314
  • Cas Number : 382-43-4
  • Molecular Weight : 158.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4244

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4244
Availability details for
Quantity
  • Availability
  • Add to Cart