COMH42122635 Combi-Blocks, Inc.

potassium dihydrate oxoborinate

  • MDL number MFCD01114230
  • Cas Number : 162379-51-3
  • Molecular Weight : 117.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4429

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4429
Availability details for
Quantity
  • Availability
  • Add to Cart