COMH4212263A Combi-Blocks, Inc.

3-Amino-3-(2,4-dimethoxyphenyl)propanoic acid

  • MDL number MFCD03002434
  • Cas Number : 34841-02-6
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4586

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4586
Availability details for
Quantity
  • Availability
  • Add to Cart