COMH4212264C Combi-Blocks, Inc.

4-Bromo-2-fluoro-5-methoxybenzoic acid

  • MDL number MFCD29931556
  • Cas Number : 1620677-51-1
  • Molecular Weight : 249.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5175

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5175
Availability details for
Quantity
  • Availability
  • Add to Cart