COMH4212265C Combi-Blocks, Inc.

Endo-n-benzyl-endo-3-aminotropane

  • MDL number MFCD09839704
  • Cas Number : 101353-61-1
  • Molecular Weight : 230.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5681
Availability details for
Quantity
  • Availability
  • Add to Cart