COMH42122664 Combi-Blocks, Inc.

chloro(3-methylbutoxy)methanone

  • MDL number MFCD07776970
  • Cas Number : 628-50-2
  • Molecular Weight : 150.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5869

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5869
Availability details for
Quantity
  • Availability
  • Add to Cart