COMH42122678 Combi-Blocks, Inc.

2-Chloro-6-fluoro-4-(trifluoromethyl)aniline

  • MDL number MFCD11036237
  • Cas Number : 123973-31-9
  • Molecular Weight : 213.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6007
Availability details for
Quantity
  • Availability
  • Add to Cart