COMH4212267E Combi-Blocks, Inc.

N-(2,2-Dimethoxyethyl)-2-methylpropenamide

  • MDL number MFCD13152039
  • Cas Number : 95984-11-5
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6148
Availability details for
Quantity
  • Availability
  • Add to Cart