COMH421226A4 Combi-Blocks, Inc.

4-Benzyloxy-1-butene

  • MDL number MFCD27927634
  • Cas Number : 70388-33-9
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7278

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7278
Availability details for
Quantity
  • Availability
  • Add to Cart