COMH421226A6 Combi-Blocks, Inc.

9,10-Dihydro-9,10-ethanoanthracene

  • MDL number MFCD00204036
  • Cas Number : 5675-64-9
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7319

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-7319
Availability details for
Quantity
  • Availability
  • Add to Cart