COMH421226E8 Combi-Blocks, Inc.

Poly(1,2-dihydro-2,2,4-trimethylquinoline)

  • MDL number MFCD00238760
  • Cas Number : 26780-96-1
  • Molecular Weight : 519.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8910
Availability details for
Quantity
  • Availability
  • Add to Cart