COMH421226FC Combi-Blocks, Inc.

(1R,4S,5S)-Rel-2-(tert-butoxycarbonyl)-2-azabicyclo[2.1.1]hexane-5-carboxylic acid

  • MDL number MFCD22209378
  • Cas Number : 615575-74-1
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9635
Availability details for
Quantity
  • Availability
  • Add to Cart